Product Details

SNP ID
rs12081155
Assay Type
Functionally Tested
NCBI dbSNP Submissions
12
Location
Chr.1:169110252 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTTGTGTACCCACTCCCTCCCTGG[C/G]CCCCTCCTACCACATATACTTCTTT
Phenotype
MIM: 182330
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
ATP1B1 PubMed Links

Gene Details

Gene
ATP1B1
Gene Name
ATPase Na+/K+ transporting subunit beta 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001677.3 Intron NP_001668.1
Gene
LOC101928596
Gene Name
uncharacterized LOC101928596
There are no transcripts associated with this gene.

View Full Product Details