Product Details
- SNP ID
-
rs12063398
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
25
- Location
-
Chr.1:201284851 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTTCCAAGCTGGATCCCGGGCCTG[A/G]CACCCCCTCCTCCAGCCTCTGAGAG
- Phenotype
-
MIM: 601975
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PKP1
PubMed Links
Gene Details
- Gene
- PKP1
- Gene Name
- plakophilin 1
View Full Product Details