Product Details

SNP ID
rs3116073
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:100135927 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCAAATATGAGACTATTTCTTTT[C/T]TTTTTTTTTTTTTTGAGAATAGAAC
Phenotype
MIM: 609110
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FBXO43 PubMed Links

Gene Details

Gene
FBXO43
Gene Name
F-box protein 43
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001029860.3 Intron NP_001025031.2
XM_011516987.2 Intron XP_011515289.1
XM_011516989.2 Intron XP_011515291.1
XM_011516990.1 Intron XP_011515292.1
XM_011516992.2 Intron XP_011515294.1
XM_017013358.1 Intron XP_016868847.1

View Full Product Details