Product Details

SNP ID
rs12140831
Assay Type
Functionally tested
NCBI dbSNP Submissions
33
Location
Chr.1:212847379 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCTTTTGCGCCATCCATATTGCTT[C/T]TGAGCCATTGTAAAGTCGAAAAATT
Phenotype
MIM: 610864
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FLVCR1-AS1 PubMed Links

Gene Details

Gene
FLVCR1-AS1
Gene Name
FLVCR1 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

Gene
SPATA45
Gene Name
spermatogenesis associated 45
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024601.2 Intron NP_001019772.1

View Full Product Details