Product Details

SNP ID
rs11119975
Assay Type
Functionally tested
NCBI dbSNP Submissions
25
Location
Chr.1:212574380 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTATTATTAATAGCTAGTTATACTG[C/T]ACTGTGATCAAATAATCAAGCTTAT
Phenotype
MIM: 603148
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATF3 PubMed Links

Gene Details

Gene
ATF3
Gene Name
activating transcription factor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001030287.3 Intron NP_001025458.1
NM_001040619.2 Intron NP_001035709.1
NM_001206484.2 Intron NP_001193413.2
NM_001206486.2 Intron NP_001193415.1
NM_001206488.2 Intron NP_001193417.2
NM_001674.3 Intron NP_001665.1
XM_005273146.1 Intron XP_005273203.1
XM_011509579.1 Intron XP_011507881.1

View Full Product Details