Product Details

SNP ID
rs7208440
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:47825646 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAATCATGGAATGGAGGGAAGTAAC[A/G]GGGAACTATTGTTTAATGGGTGTAG
Phenotype
MIM: 611825 MIM: 606735
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LRRC46 PubMed Links

Gene Details

Gene
LRRC46
Gene Name
leucine rich repeat containing 46
There are no transcripts associated with this gene.

Gene
MRPL10
Gene Name
mitochondrial ribosomal protein L10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145255.3 Intron NP_660298.2
NM_148887.2 Intron NP_683685.1
Gene
OSBPL7
Gene Name
oxysterol binding protein like 7
There are no transcripts associated with this gene.

View Full Product Details