Product Details
- SNP ID
-
hCV32164220
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:121762981 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGCTTAACTTGCATTCACAGCATG[A/T]CTTTATTTTCATGTGTTTTTTAGTT
- Phenotype
-
MIM: 600575
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TSN
PubMed Links
Gene Details
- Gene
- TSN
- Gene Name
- translin
There are no transcripts associated with this gene.
View Full Product Details