Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001199140.1 | Intron | NP_001186069.1 | |||
NM_031445.2 | Intron | NP_113633.2 | |||
XM_005263804.4 | Intron | XP_005263861.1 | |||
XM_005263806.4 | Intron | XP_005263863.1 | |||
XM_005263807.4 | Intron | XP_005263864.1 | |||
XM_011511952.2 | Intron | XP_011510254.1 | |||
XM_011511954.2 | Intron | XP_011510256.1 | |||
XM_011511955.2 | Intron | XP_011510257.1 | |||
XM_011511956.2 | Intron | XP_011510258.1 | |||
XM_017005063.1 | Intron | XP_016860552.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
XM_017005446.1 | Intron | XP_016860935.1 |