Product Details
- SNP ID
-
rs7194625
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:77724789 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTCCACATAGGAAGGGCACCATTTG[C/T]GGTTTTAAATCCAAATGCTTGCATA
- Phenotype
-
MIM: 609231
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
NUDT7
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs80159064] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NUDT7
- Gene Name
- nudix hydrolase 7
View Full Product Details