Product Details

SNP ID
rs12306769
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:13374948 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCTCCGGAGTCCTTTGGTGATGGA[A/G]AACAGGGCTGACAGGATTCAAATGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC01559 PubMed Links

Gene Details

Gene
LINC01559
Gene Name
long intergenic non-protein coding RNA 1559
There are no transcripts associated with this gene.

View Full Product Details