Product Details

SNP ID
rs5744046
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:12911471 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGGCCTTCTTTCCTGTGTGTCTA[C/T]TTCCAAATTCCCCTTTTCTTATAAG
Phenotype
MIM: 300366
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TLR8 PubMed Links

Gene Details

Gene
TLR8
Gene Name
toll like receptor 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016610.3 Intron NP_057694.2
NM_138636.5 Intron NP_619542.1
XM_011545529.1 Intron XP_011543831.1
XM_011545530.2 Intron XP_011543832.1
Gene
TLR8-AS1
Gene Name
TLR8 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details