Product Details

SNP ID
rs12190581
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:42159121 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGCCTCTACCAAGGATAAAGAGAT[C/T]GCCATGGGTCCTATAGCACTGATTG
Phenotype
MIM: 600364
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GUCA1A PubMed Links

Gene Details

Gene
GUCA1A
Gene Name
guanylate cyclase activator 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000409.4 Intron NP_000400.2
NM_001319061.1 Intron NP_001305990.1
NM_001319062.1 Intron NP_001305991.1
XM_006715073.3 Intron XP_006715136.1
XM_011514537.2 Intron XP_011512839.1
XM_011514539.1 Intron XP_011512841.1

View Full Product Details