Product Details

SNP ID
rs3793023
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:43015021 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTGTTTATATGGCAGAGAAACTAA[A/C]ATTTTTTTTCTCCACCTTTGGCTGG
Phenotype
MIM: 611248 MIM: 143170 MIM: 601646 MIM: 613475
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
KLHDC3 PubMed Links

Gene Details

Gene
KLHDC3
Gene Name
kelch domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_057161.3 Intron NP_476502.1
Gene
MEA1
Gene Name
male-enhanced antigen 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318942.1 Intron NP_001305871.1
NM_001318943.1 Intron NP_001305872.1
NM_014623.3 Intron NP_055438.1
XM_005249121.3 Intron XP_005249178.1
XM_017010868.1 Intron XP_016866357.1
Gene
PPP2R5D
Gene Name
protein phosphatase 2 regulatory subunit B'delta
There are no transcripts associated with this gene.

Gene
RRP36
Gene Name
ribosomal RNA processing 36
There are no transcripts associated with this gene.

View Full Product Details