Product Details

SNP ID
rs7225330
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:16678385 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAGATCATAATAGTAGCTTAAAAC[A/G]ATAGCCAAGGCAGTTAGAGTTGTGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC144A PubMed Links

Gene Details

Gene
CCDC144A
Gene Name
coiled-coil domain containing 144A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014695.2 Intron NP_055510.1
XM_011524089.2 Intron XP_011522391.1
XM_017025429.1 Intron XP_016880918.1
XM_017025430.1 Intron XP_016880919.1

View Full Product Details