Product Details

SNP ID
rs11133443
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:56470143 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAAAAAATATGTATTGAAATTAAG[G/T]TTGAAGCATATTAATGGGATACTGG
Phenotype
MIM: 172439 MIM: 602122
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PAICS PubMed Links

Gene Details

Gene
PAICS
Gene Name
phosphoribosylaminoimidazole carboxylase; phosphoribosylaminoimidazolesuccinocarboxamide synthase
There are no transcripts associated with this gene.

Gene
SRP72
Gene Name
signal recognition particle 72
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267722.1 Intron NP_001254651.1
NM_006947.3 Intron NP_008878.3
XM_005265765.4 Intron XP_005265822.3

View Full Product Details