Product Details
- SNP ID
-
rs4975161
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:79177271 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TATCTTTTTAGTTGAATTTTCTCTT[C/T]CCTTTGGGAGCATCTGAACCAACAA
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LINC01088
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77422609] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LINC01088
- Gene Name
- long intergenic non-protein coding RNA 1088
There are no transcripts associated with this gene.
- Gene
- NAA11
- Gene Name
- N(alpha)-acetyltransferase 11, NatA catalytic subunit
View Full Product Details