Product Details

SNP ID
rs12724404
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:47417174 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGGCCCGCTGCCCGCTGAGCCCCTC[A/C]TGGCCTTGGCCGGGCCGGCAGCCGC
Phenotype
MIM: 601094
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FOXE3 PubMed Links

Gene Details

Gene
FOXE3
Gene Name
forkhead box E3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012186.2 1103 Missense Mutation ATG,CTG M287L NP_036318.1
Gene
LINC01389
Gene Name
long intergenic non-protein coding RNA 1389
There are no transcripts associated with this gene.

View Full Product Details