Product Details

SNP ID
rs7703456
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:10357260 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGAAAAGCAGTACAATATTGGAATA[C/G]ATGTTGCAAGTGTTGATGTGCAGTT
Phenotype
MIM: 613297
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC101929977 PubMed Links

Gene Details

Gene
LOC101929977
Gene Name
uncharacterized LOC101929977
There are no transcripts associated with this gene.

Gene
MARCH6
Gene Name
membrane associated ring-CH-type finger 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270660.1 Intron NP_001257589.1
NM_001270661.1 Intron NP_001257590.1
NM_005885.3 Intron NP_005876.2
XM_011513932.2 Intron XP_011512234.1
XM_011513934.1 Intron XP_011512236.1
XM_011513936.2 Intron XP_011512238.1
XM_011513937.2 Intron XP_011512239.1
XM_017008944.1 Intron XP_016864433.1

View Full Product Details