Product Details
- SNP ID
-
rs11574994
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:46056479 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CACTTATCTGGAACTTTTCGGAAAT[A/T]GGAACTAGCATCATGGTATACACTG
- Phenotype
-
MIM: 603101
MIM: 616453
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CPB2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs548262831] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CPB2
- Gene Name
- carboxypeptidase B2
- Gene
- CPB2-AS1
- Gene Name
- CPB2 antisense RNA 1
There are no transcripts associated with this gene.
- Gene
- ZC3H13
- Gene Name
- zinc finger CCCH-type containing 13
There are no transcripts associated with this gene.
View Full Product Details