Product Details

SNP ID
rs12208297
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:125994603 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGTAAATATTGGAAGAAAAAAAA[C/T]GGGGTATATACCCAAAGTAATATAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TRMT11 PubMed Links

Gene Details

Gene
TRMT11
Gene Name
tRNA methyltransferase 11 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031712.2 Intron NP_001026882.2
XM_017011168.1 Intron XP_016866657.1
XM_017011169.1 Intron XP_016866658.1

View Full Product Details