Product Details
- SNP ID
-
rs12333055
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:112322188 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTCTTGAATAGGGGTGTTGGAAAAT[A/G]TGGAACAGGGAGAAAAACCTCACCA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RFPL4B
PubMed Links
Gene Details
- Gene
- RFPL4B
- Gene Name
- ret finger protein like 4B
View Full Product Details