Product Details

SNP ID
rs12427768
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:102789381 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTTAGTAAGTGTTATTAAACAACA[A/G]GTTGATCTAGAGCTAAATGCTCATT
Phenotype
MIM: 611613
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BIVM PubMed Links

Gene Details

Gene
BIVM
Gene Name
basic, immunoglobulin-like variable motif containing
There are no transcripts associated with this gene.

Gene
KDELC1
Gene Name
KDEL motif containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318732.1 Intron NP_001305661.1
NM_024089.2 Intron NP_076994.2
XM_005254075.2 Intron XP_005254132.1

View Full Product Details