Product Details
- SNP ID
-
rs12868192
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:101054021 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAAAATAGATTATAAAACAACCTGG[A/T]GGTCACAGGATTCTGAGATAATCCC
- Phenotype
-
MIM: 611549
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
NALCN
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs145134837] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NALCN
- Gene Name
- sodium leak channel, non-selective
- Gene
- NALCN-AS1
- Gene Name
- NALCN antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details