Product Details

SNP ID
rs12053674
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:30747177 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAGCCATATCCACAGCCCCCACAG[C/G]AGTTTCTGTAGTAGTTGCAACACAT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
KRTAP21-1 PubMed Links

Gene Details

Gene
KRTAP21-1
Gene Name
keratin associated protein 21-1
There are no transcripts associated with this gene.

Gene
KRTAP21-2
Gene Name
keratin associated protein 21-2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181617.1 26 Missense Mutation TCC,TGC S9C NP_853648.1

View Full Product Details