Product Details

SNP ID
rs11701787
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:29200534 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGATTTTGAGAAGGATTGAGTTGA[A/G]TATGTGGATTAATTTGTGAAATATT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC00189 PubMed Links

Gene Details

Gene
LINC00189
Gene Name
long intergenic non-protein coding RNA 189
There are no transcripts associated with this gene.

View Full Product Details