Product Details

SNP ID
rs12151388
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:176279301 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACATCATATTTTATTACCTGGTCA[A/G]TTTAGAGCACTAATCAGAACTTTAT
Phenotype
MIM: 608555
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MTX2 PubMed Links

Gene Details

Gene
MTX2
Gene Name
metaxin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001006635.2 Intron NP_001006636.1
NM_001319097.1 Intron NP_001306026.1
NM_001319098.1 Intron NP_001306027.1
NM_006554.4 Intron NP_006545.1
XM_011510497.2 Intron XP_011508799.1

View Full Product Details