Product Details

SNP ID
rs11798681
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:132630365 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCACAGGGAACCTGCTCAGCCACC[A/T]GCTTAACTGAATATTGGCTGATGTT
Phenotype
MIM: 300545
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
HS6ST2 PubMed Links

Gene Details

Gene
HS6ST2
Gene Name
heparan sulfate 6-O-sulfotransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077188.1 Intron NP_001070656.1
NM_147175.3 Intron NP_671704.3
XM_005262490.3 Intron XP_005262547.1
XM_005262491.2 Intron XP_005262548.1
XM_011531406.1 Intron XP_011529708.1
XM_011531407.2 Intron XP_011529709.1
XM_011531408.2 Intron XP_011529710.1
XM_017029944.1 Intron XP_016885433.1
XM_017029945.1 Intron XP_016885434.1
XM_017029946.1 Intron XP_016885435.1

View Full Product Details