Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001184896.1 | 4743 | UTR 3 | NP_001171825.1 | ||
NM_001184897.1 | 4743 | UTR 3 | NP_001171826.1 | ||
NM_001184898.1 | 4743 | Intron | NP_001171827.1 | ||
NM_015107.2 | 4743 | UTR 3 | NP_055922.1 | ||
XM_005261996.1 | 4743 | UTR 3 | XP_005262053.1 | ||
XM_005261997.3 | 4743 | UTR 3 | XP_005262054.1 | ||
XM_005261999.1 | 4743 | UTR 3 | XP_005262056.1 | ||
XM_005262000.1 | 4743 | UTR 3 | XP_005262057.1 | ||
XM_011530778.1 | 4743 | UTR 3 | XP_011529080.1 | ||
XM_017029361.1 | 4743 | UTR 3 | XP_016884850.1 | ||
XM_017029362.1 | 4743 | UTR 3 | XP_016884851.1 |