Product Details

SNP ID
rs4366064
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:124477973 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTAAAAAAGAATCCTTAAATGAC[A/C]AACTGAGATAAAATACTTCTATCAT
Phenotype
MIM: 603046
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
RNF139 PubMed Links

Gene Details

Gene
RNF139
Gene Name
ring finger protein 139
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007218.3 Intron NP_009149.2
XM_017013002.1 Intron XP_016868491.1
Gene
RNF139-AS1
Gene Name
RNF139 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details