Product Details

SNP ID
rs4253814
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:186266243 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGGATGAAGACCATTTTGGAGGAA[A/G]AAAAGCACCCTTATTAAGAATTGCA
Phenotype
MIM: 264900 MIM: 229000
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
F11 PubMed Links

Gene Details

Gene
F11
Gene Name
coagulation factor XI
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000128.3 299 UTR 5 NP_000119.1
XM_005262821.3 299 UTR 5 XP_005262878.1
XM_005262822.3 299 UTR 5 XP_005262879.1
XM_005262823.3 299 UTR 5 XP_005262880.1
XM_006714137.2 299 UTR 5 XP_006714200.1
XM_017007884.1 299 UTR 5 XP_016863373.1
XM_017007885.1 299 UTR 5 XP_016863374.1
XM_017007886.1 299 UTR 5 XP_016863375.1
Gene
KLKB1
Gene Name
kallikrein B1
There are no transcripts associated with this gene.

View Full Product Details