Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001114182.2 | 665 | Intron | NP_001107654.1 | ||
NM_001145256.1 | 665 | Intron | NP_001138728.1 | ||
NM_001145257.1 | 665 | Intron | NP_001138729.1 | ||
NM_001145258.1 | 665 | Intron | NP_001138730.1 | ||
NM_016123.3 | 665 | Intron | NP_057207.2 | ||
XM_005268943.3 | 665 | Intron | XP_005269000.1 | ||
XM_005268944.4 | 665 | Intron | XP_005269001.1 | ||
XM_005268945.4 | 665 | Intron | XP_005269002.1 | ||
XM_005268947.4 | 665 | Intron | XP_005269004.1 | ||
XM_005268948.2 | 665 | Intron | XP_005269005.1 | ||
XM_005268949.2 | 665 | Intron | XP_005269006.1 | ||
XM_006719438.3 | 665 | Intron | XP_006719501.1 | ||
XM_006719439.2 | 665 | Intron | XP_006719502.1 | ||
XM_006719440.2 | 665 | Intron | XP_006719503.1 | ||
XM_006719441.2 | 665 | Intron | XP_006719504.1 | ||
XM_006719442.2 | 665 | Intron | XP_006719505.1 | ||
XM_006719443.3 | 665 | Intron | XP_006719506.1 | ||
XM_011538431.2 | 665 | Intron | XP_011536733.1 | ||
XM_011538432.1 | 665 | Intron | XP_011536734.1 | ||
XM_011538433.2 | 665 | UTR 5 | XP_011536735.1 | ||
XM_017019390.1 | 665 | Intron | XP_016874879.1 | ||
XM_017019391.1 | 665 | Intron | XP_016874880.1 |