Product Details
- SNP ID
-
rs4251445
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:43764946 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GAGCATCCCAAAAACCTGCAGCCAT[A/G]CCCTTGCTTTTGACTGTCTGCTTTT
- Phenotype
-
MIM: 606883
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
IRAK4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs138653083] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- IRAK4
- Gene Name
- interleukin 1 receptor associated kinase 4
- Gene
- PUS7L
- Gene Name
- pseudouridylate synthase 7 like
There are no transcripts associated with this gene.
View Full Product Details