Product Details

SNP ID
rs4240234
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:16551007 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTAGAATCTGTAGGTTCAAAAAG[G/T]TCTTGAAGACCACCTAGCATATTTC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FAM49A PubMed Links

Gene Details

Gene
FAM49A
Gene Name
family with sequence similarity 49 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030797.3 3154 UTR 3 NP_110424.1
XM_005262630.3 3154 UTR 3 XP_005262687.1
XM_006712108.3 3154 UTR 3 XP_006712171.1

View Full Product Details