Product Details

SNP ID
rs3936007
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:29930664 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGAACTGGGCTAGACCAGTTTACT[C/G]CCTTCACCCCGGCTCTCGGGGGCTT
Phenotype
MIM: 606779
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
EFCAB5 PubMed Links

Gene Details

Gene
EFCAB5
Gene Name
EF-hand calcium binding domain 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145053.1 Intron NP_001138525.1
NM_198529.3 Intron NP_940931.2
XM_011524759.1 Intron XP_011523061.1
XM_011524763.2 Intron XP_011523065.1
XM_011524765.1 Intron XP_011523067.1
XM_011524766.2 Intron XP_011523068.1
XM_011524768.2 Intron XP_011523070.1
XM_017024591.1 Intron XP_016880080.1
XM_017024592.1 Intron XP_016880081.1
Gene
SSH2
Gene Name
slingshot protein phosphatase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282129.1 Intron NP_001269058.1
NM_001282130.1 Intron NP_001269059.1
NM_001282131.1 Intron NP_001269060.1
NM_033389.3 Intron NP_203747.2
XM_005258058.3 Intron XP_005258115.1
XM_005258059.2 Intron XP_005258116.1
XM_005258060.3 Intron XP_005258117.1
XM_006722149.3 Intron XP_006722212.1
XM_011525402.2 Intron XP_011523704.1
XM_011525404.2 Intron XP_011523706.1
XM_011525405.2 Intron XP_011523707.1
XM_011525406.2 Intron XP_011523708.1
XM_011525407.1 Intron XP_011523709.1
XM_017025251.1 Intron XP_016880740.1
XM_017025252.1 Intron XP_016880741.1
XM_017025253.1 Intron XP_016880742.1
XM_017025254.1 Intron XP_016880743.1
XM_017025255.1 Intron XP_016880744.1

View Full Product Details