Product Details

SNP ID
rs3111564
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:8510881 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGCGTCGGGGCGCGGCGGCCCGG[C/G]CGCGGGGGCCGCGGGGGCGCCGGGG
Phenotype
MIM: 601480 MIM: 606466
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MYO1F PubMed Links

Gene Details

Gene
MYO1F
Gene Name
myosin IF
There are no transcripts associated with this gene.

Gene
PRAM1
Gene Name
PML-RARA regulated adaptor molecule 1
There are no transcripts associated with this gene.

Gene
ZNF414
Gene Name
zinc finger protein 414
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146175.1 1187 Missense Mutation CCC,GCC P357A NP_001139647.1
NM_032370.2 1187 Intron NP_115746.2

View Full Product Details