Product Details

SNP ID
rs3823720
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:41961573 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATAAAACAAAAATGGTTAGAAAAC[A/G]TTTAACACAGACATTAAGCAGGAGT
Phenotype
MIM: 165240
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GLI3 PubMed Links

Gene Details

Gene
GLI3
Gene Name
GLI family zinc finger 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000168.5 7591 UTR 3 NP_000159.3
XM_011515274.2 7591 Intron XP_011513576.1
XM_017011997.1 7591 Intron XP_016867486.1

View Full Product Details