Product Details

SNP ID
rs3219174
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:201700958 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAAGTATAGGGAAGAACAACACCAG[A/G]CTGGGGATCGATGCAGTTTTTTGCT
Phenotype
MIM: 606352 MIM: 606575
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ALS2 PubMed Links

Gene Details

Gene
ALS2
Gene Name
ALS2, alsin Rho guanine nucleotide exchange factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135745.1 5515 Intron NP_001129217.1
NM_020919.3 5515 UTR 3 NP_065970.2
XM_006712654.2 5515 UTR 3 XP_006712717.1
XM_006712655.3 5515 UTR 3 XP_006712718.1
XM_011511530.2 5515 UTR 3 XP_011509832.1
XM_017004569.1 5515 UTR 3 XP_016860058.1
XM_017004570.1 5515 Intron XP_016860059.1
XM_017004571.1 5515 UTR 3 XP_016860060.1
XM_017004572.1 5515 UTR 3 XP_016860061.1
Gene
MPP4
Gene Name
membrane palmitoylated protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033066.2 5515 Intron NP_149055.1
XM_017004620.1 5515 Intron XP_016860109.1

View Full Product Details