Product Details

SNP ID
rs7940905
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:73208922 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTTCAGGAGGCACTTAGGATTAGA[C/T]AAAGTCATCAAGGTGGGGCCCACAT
Phenotype
MIM: 600041
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
P2RY2 PubMed Links

Gene Details

Gene
P2RY2
Gene Name
purinergic receptor P2Y2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002564.3 Intron NP_002555.3
NM_176071.2 Intron NP_788085.2
NM_176072.2 Intron NP_788086.2
XM_005274019.4 Intron XP_005274076.1
XM_005274020.4 Intron XP_005274077.1
XM_005274021.4 Intron XP_005274078.1
XM_011545074.2 Intron XP_011543376.1
XM_017017839.1 Intron XP_016873328.1

View Full Product Details