Product Details

SNP ID
rs7941549
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:11846741 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACAACTTATTTTTATTTCTATTGTC[C/T]GACTTCTTATAAAATCCATTCATCA
Phenotype
MIM: 614460
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
USP47 PubMed Links

Gene Details

Gene
USP47
Gene Name
ubiquitin specific peptidase 47
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282659.1 Intron NP_001269588.1
NM_017944.3 Intron NP_060414.3
XM_005252997.1 Intron XP_005253054.1
XM_011520196.1 Intron XP_011518498.1
XM_017017953.1 Intron XP_016873442.1
XM_017017954.1 Intron XP_016873443.1

View Full Product Details