Product Details

SNP ID
rs104893847
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:67740508 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAGAAAAATATCCATAGATAAGT[G/A]GATCAAAGCATGGGTTTAAAAAGGC
Phenotype
MIM: 138850
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
GNRHR PubMed Links

Gene Details

Gene
GNRHR
Gene Name
gonadotropin releasing hormone receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000406.2 2710 Missense Mutation CCA,CTA P320L NP_000397.1
NM_001012763.1 2710 UTR 3 NP_001012781.1

View Full Product Details