Product Details

SNP ID
rs4147652
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:8311493 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGGCAAAGTAGTCGGGTGGCCGT[A/G]AGGGTGCAGTGTCACAGGTAAGGCT
Phenotype
MIM: 606475 MIM: 602139
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CD320 PubMed Links

Gene Details

Gene
CD320
Gene Name
CD320 molecule
There are no transcripts associated with this gene.

Gene
NDUFA7
Gene Name
NADH:ubiquinone oxidoreductase subunit A7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005001.4 409 UTR 3 NP_004992.2

View Full Product Details