Product Details

SNP ID
rs7737266
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:95565048 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTCAAGGATTTAGATATATTCTAT[A/G]TTTTGGGTTGGCTCATGTTCTAAAC
Phenotype
MIM: 610011
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARSK PubMed Links

Gene Details

Gene
ARSK
Gene Name
arylsulfatase family member K
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198150.2 Intron NP_937793.1
XM_005271904.3 Intron XP_005271961.1

View Full Product Details