Product Details

SNP ID
rs4253233
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:49458697 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTTTAATTCTGAGGTAGACATCA[G/T]GCAAACAAACATCAAGTGCAGCCAA
Phenotype
MIM: 609413
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ERCC6 PubMed Links

Gene Details

Gene
ERCC6
Gene Name
ERCC excision repair 6, chromatin remodeling factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000124.3 4776 UTR 3 NP_000115.1
NM_001277058.1 4776 Intron NP_001263987.1
NM_001277059.1 4776 Intron NP_001263988.1

View Full Product Details