Product Details

SNP ID
rs3219287
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:50384466 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGGCTTGAGCAGCCAGGCACATAG[A/G]TCTGAGAGACGGTTGCGAGGGCACC
Phenotype
MIM: 600380 MIM: 174761
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NR1H2 PubMed Links

Gene Details

Gene
NR1H2
Gene Name
nuclear receptor subfamily 1 group H member 2
There are no transcripts associated with this gene.

Gene
POLD1
Gene Name
DAN polymerase delta 1, catalytic subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256849.1 Intron NP_001243778.1
NM_001308632.1 Intron NP_001295561.1
NM_002691.3 Intron NP_002682.2
XM_005259008.4 Intron XP_005259065.1
XM_011527038.1 Intron XP_011525340.1
XM_017026881.1 Intron XP_016882370.1
XM_017026882.1 Intron XP_016882371.1

View Full Product Details