Product Details

SNP ID
rs4924857
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:17479901 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCATCTACCAACTCCTAAGCACCTG[G/T]TTTGTTAACCATGGCCTATCCCCAG
Phenotype
MIM: 609878
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MED9 PubMed Links

Gene Details

Gene
MED9
Gene Name
mediator complex subunit 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018019.2 Intron NP_060489.1

View Full Product Details