Product Details

SNP ID
rs3211635
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:57748085 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCAATTTCAGTTCCTTTCTAAGC[T/G]TTGTCAGTCAAGGGGCTCCACTGAC
Phenotype
MIM: 605476 MIM: 123829 MIM: 613336 MIM: 181035
Polymorphism
T/G, Transversion Substitution
Allele Nomenclature
Literature Links
AGAP2 PubMed Links
Additional Information
For this assay, SNP(s) [rs3472] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
AGAP2
Gene Name
ArfGAP with GTPase domain, ankyrin repeat and PH domain 2
There are no transcripts associated with this gene.

Gene
CDK4
Gene Name
cyclin dependent kinase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000075.3 1644 UTR 3 NP_000066.1
Gene
MARCH9
Gene Name
membrane associated ring-CH-type finger 9
There are no transcripts associated with this gene.

Gene
MIR6759
Gene Name
microRNA 6759
There are no transcripts associated with this gene.

Gene
TSPAN31
Gene Name
tetraspanin 31
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005981.3 1644 UTR 3 NP_005972.1
XM_005269074.3 1644 UTR 3 XP_005269131.2
XM_017019793.1 1644 UTR 3 XP_016875282.1

View Full Product Details