Product Details

SNP ID
rs4252548
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:55368304 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTCAGGGAAGAGCCACCTGCCCGG[C/T]GCAGCCACTGCACGTGCCGCAGGTA
Phenotype
MIM: 147681
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
FAM71E2 PubMed Links

Gene Details

Gene
FAM71E2
Gene Name
family with sequence similarity 71 member E2
There are no transcripts associated with this gene.

Gene
IL11
Gene Name
interleukin 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000641.3 488 Missense Mutation CAC,CGC H112R NP_000632.1
NM_001267718.1 488 Missense Mutation CAC,CGC H33R NP_001254647.1
Gene
TMEM190
Gene Name
transmembrane protein 190
There are no transcripts associated with this gene.

View Full Product Details