Product Details

SNP ID
rs4586622
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:224472838 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAACAGAAACCATCTTCTGAGACA[C/T]ACATTCTAATAAATGTATCAACAGT
Phenotype
MIM: 603136
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CUL3 PubMed Links

Gene Details

Gene
CUL3
Gene Name
cullin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001257197.1 4102 UTR 3 NP_001244126.1
NM_001257198.1 4102 UTR 3 NP_001244127.1
NM_003590.4 4102 UTR 3 NP_003581.1

View Full Product Details