Product Details

SNP ID
rs6673915
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:108694369 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTAATAGAGGAAAGTCTCGTTTTT[A/T]AGTTTCAAATGTGTTTTGAAACAAG
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PRPF38B PubMed Links

Gene Details

Gene
PRPF38B
Gene Name
pre-mRNA processing factor 38B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018061.2 Intron NP_060531.2
XM_011541687.2 Intron XP_011539989.1
XM_017001596.1 Intron XP_016857085.1
XM_017001597.1 Intron XP_016857086.1
XM_017001598.1 Intron XP_016857087.1

View Full Product Details