Product Details

SNP ID
rs3218326
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:37128052 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTCAACAGGGTCCTTCTGAGGCTC[A/G]GCGCAGAGCAGGCAGCTGCCTGCCT
Phenotype
MIM: 146710
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IL2RB PubMed Links

Gene Details

Gene
IL2RB
Gene Name
interleukin 2 receptor subunit beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000878.3 1831 UTR 3 NP_000869.1

View Full Product Details